CAS RN 11110-52-4
IUPAC Name:
mercury sodium
Synonyms:
mercury sodium
Molecular Weight (g/mol):
223.58
Molecular Formula:
HgNa
InChi Key:
MJGFBOZCAJSGQW-UHFFFAOYSA-N
SMILES:
[Na].[Hg]
1
–
3
of
3
results
1
Promotions Available
Sodium mercury amalgam, ca. 5% sodium
Sodium mercury amalgam, ca. 5% sodium
3
Sodium mercury amalgam, ca. 20% sodium
CAS: 11110-52-4 Molecular Formula: HgNa Molecular Weight (g/mol): 223.582 MDL Number: MFCD00210362 InChI Key: MJGFBOZCAJSGQW-UHFFFAOYSA-N Synonym: sodium amalgam,sodium mercury amalgam,sodium mercury,sodium-amalgam,mercury sodium,sodium mercury amalgam, beads, na 10 %,sodium mercury amalgam, beads trace metals basis, na 5 %,sodium mercury amalgam, beads trace metals basis, na 20 % PubChem CID: 9837419 IUPAC Name: mercury;sodium SMILES: [Na].[Hg]