accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNON

CAS RN 111249-50-4

IUPAC Name: 3-(3-methylphenyl)-5-phenyl-1,2,4-oxadiazole
Synonyms: 3-(3-methylphenyl)-5-phenyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 236.27
Molecular Formula: C15H12N2O
InChi Key: LVOLWXPQHWLZBL-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=NOC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results