accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3HNOSPOOH3COH3C

CAS RN 1113-02-6

IUPAC Name: dimethyl {[(methylcarbamoyl)methyl]sulfanyl}phosphonate
Synonyms: omethoate folimat O-analog of dimethoate dimethoate O-analog dimethoate-met PO-dimethoate omethoat dimethoate PO isologue dimethoate oxygen analog dimethoxon
Molecular Weight (g/mol): 213.19
Molecular Formula: C5H12NO4PS
InChi Key: PZXOQEXFMJCDPG-UHFFFAOYSA-N
SMILES: CNC(=O)CSP(=O)(OC)OC
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results