accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3CH3H3COHCH2(E)(E)

CAS RN 1113-21-9

IUPAC Name: (6E,10E)-3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
Synonyms: geranyl linalool
Molecular Weight (g/mol): 290.49
Molecular Formula: C20H34O
InChi Key: IQDXAJNQKSIPGB-HQSZAHFGNA-N
SMILES: CC(C)=CCC\C(C)=C\CC\C(C)=C\CCC(C)(O)C=C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)