CAS RN 1113-68-4
IUPAC Name:
N-acetyl-N-methylacetamide
Synonyms:
acetamide, N-acetyl-N-methyl-
Molecular Weight (g/mol):
115.13
Molecular Formula:
C5H9NO2
InChi Key:
ZNQFZPCFVNOXJQ-UHFFFAOYSA-N
SMILES:
CN(C(C)=O)C(C)=O
1
–
1
of
1
results
1
N-Methyldiacetamide, 90+%
CAS: 1113-68-4 Molecular Formula: C5H9NO2 Molecular Weight (g/mol): 115.132 MDL Number: MFCD00014968 InChI Key: ZNQFZPCFVNOXJQ-UHFFFAOYSA-N Synonym: n-methyldiacetamide,acetamide, n-acetyl-n-methyl,diacetamide, n-methyl,unii-767ut09b3u,n,n-diacetylmethylamine,n-methyl diacetamide,methyldiacetylamine,nn-diacetylmethylamine,acmc-1btf5,n-methyl-n-acetylacetamide PubChem CID: 14214 IUPAC Name: N-acetyl-N-methylacetamide SMILES: CC(=O)N(C)C(=O)C