accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)HO(S)OH(R)HO(S)NHOHO(R)HOOHOO

CAS RN 1113-83-3

IUPAC Name: (3R,4S,5R,6S,7R)-3,5,6,7,8-pentahydroxy-4-(2-hydroxyacetamido)-2-oxooctanoic acid
Synonyms: N-glycolylneuraminic acid
Molecular Weight (g/mol): 311.24
Molecular Formula: C10H17NO10
InChi Key: YIICUBSXBNCXHW-BOSCHMPASA-N
SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](NC(=O)CO)[C@@H](O)C(=O)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results