accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

AlSi

CAS RN 11145-30-5

IUPAC Name: aluminium silicon
Synonyms: aluminium silicon
Molecular Weight (g/mol): 55.07
Molecular Formula: AlSi
InChi Key: CSDREXVUYHZDNP-UHFFFAOYSA-N
SMILES: [Al].[Si]
Form 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results