accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOCH3Br(E/Z)

CAS RN 1116-50-3

IUPAC Name: ethyl 4-bromo-3-ethoxybut-2-enoate
Synonyms: ethyl 4-bromo-3-ethoxybut-2-enoate
Molecular Weight (g/mol): 237.09
Molecular Formula: C8H13BrO3
InChi Key: FYYMWILFRUUBCX-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C(CBr)OCC
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results