accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3CH3

CAS RN 1116-76-3

IUPAC Name: trioctylamine
Synonyms: tri(N-octyl)amine
Molecular Weight (g/mol): 353.68
Molecular Formula: C24H51N
InChi Key: XTAZYLNFDRKIHJ-UHFFFAOYSA-N
SMILES: CCCCCCCCN(CCCCCCCC)CCCCCCCC
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)