accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1118-46-3

IUPAC Name: butyltin trichloride
Synonyms: butyltin trichloride
Molecular Weight (g/mol): 288.22
Molecular Formula: C4H15Cl3Sn
InChi Key: FMOHJFNRZRLZBY-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.CCCC[SnH3]
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results