accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3SiH3CCH3OOOSiH3CCH3H3CCH3CH3O

CAS RN 111830-58-1

IUPAC Name: {[3-(benzyloxy)-2-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydro-2H-pyran-4-yl]oxy}(tert-butyl)dimethylsilane
Synonyms: {[3-(benzyloxy)-2-{[(tert-butyldimethylsilyl)oxy]methyl}-3,4-dihydro-2H-pyran-4-yl]oxy}(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 464.79
Molecular Formula: C25H44O4Si2
InChi Key: CZGTZYNJABAJPH-UHFFFAOYNA-N
SMILES: CC(C)(C)[Si](C)(C)OCC1OC=CC(O[Si](C)(C)C(C)(C)C)C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results