accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClNH2(S)NH2NNH2OHOH

CAS RN 1119-34-2

IUPAC Name: hydrogen (2S)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Synonyms: arginine hcl
Molecular Weight (g/mol): 210.66
Molecular Formula: C6H15ClN4O2
InChi Key: KWTQSFXGGICVPE-WCCKRBBISA-N
SMILES: [H+].[Cl-].N[C@@H](CCCN=C(N)N)C(O)=O
Quantity 
  • (5)
  • (1)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
Molecular Weight (g/mol) 
  • (1)
  • (7)
  • (13)
  • (4)
Percent Purity 
  • (3)
  • (4)
  • (1)
  • (2)
  • (5)
  • (1)
Grade 
  • (5)
  • (1)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (9)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (5)
  • (1)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (7)
  • (13)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)