accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111900-32-4

H2N(S)(R)OHOHOO

CAS RN 111900-32-4

IUPAC Name: (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Synonyms: (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C7H11NO4
InChi Key: YFYNOWXBIBKGHB-FBCQKBJTSA-N
SMILES: N[C@]1(CC[C@H](C1)C(O)=O)C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results