accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 111964-21-7

OOCl

CAS RN 111964-21-7

IUPAC Name: 1-benzofuran-3-carbonyl chloride
Synonyms: 1-benzofuran-3-carbonyl chloride
Molecular Weight (g/mol): 180.59
Molecular Formula: C9H5ClO2
InChi Key: YUHWSRVMUQSCNL-UHFFFAOYSA-N
SMILES: ClC(=O)C1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results