accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClCH3NH3CH3CCH3

CAS RN 112-02-7

IUPAC Name: hexadecyltrimethylazanium chloride
Synonyms: cetrimonium chloride cetyl trimethyl ammonium chloride
Molecular Weight (g/mol): 320
Molecular Formula: C19H42ClN
InChi Key: WOWHHFRSBJGXCM-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (4)
  • (4)
Physical Form 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (8)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)