accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHO

CAS RN 112-05-0

IUPAC Name: nonanoic acid
Synonyms: nonanoic acid 1-octanecarboxyic acid n-pelargonate pelargonsaeure pelargon pergonic acid pelargic acid emery L-114 hexacid C-9 cirrasol 185a
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: FBUKVWPVBMHYJY-UHFFFAOYSA-N
SMILES: CCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (7)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)