accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-12-9

H3CCH3O

CAS RN 112-12-9

IUPAC Name: undecan-2-one
Synonyms: undecan-2-one caswell no. 573O mgk dog and cat repellent Mnk 2-undecanone (natural) rue ketone epa pesticide chemical code 044102
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: KYWIYKKSMDLRDC-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(C)=O
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)