accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CClO

CAS RN 112-13-0

IUPAC Name: decanoyl chloride
Synonyms: decanoyl chloride
Molecular Weight (g/mol): 190.71
Molecular Formula: C10H19ClO
InChi Key: IPIVAXLHTVNRBS-UHFFFAOYSA-N
SMILES: CCCCCCCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)