accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3O

CAS RN 112-14-1

IUPAC Name: octyl acetate
Synonyms: octyl acetate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: YLYBTZIQSIBWLI-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (5)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results