accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-17-4

IUPAC Name: decyl acetate
Synonyms: decyl acetate
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: NUPSHWCALHZGOV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCOC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results