accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 112-29-8

IUPAC Name: 1-bromodecane
Synonyms: 1-bromodecane
Molecular Weight (g/mol): 221.18
Molecular Formula: C10H21Br
InChi Key: MYMSJFSOOQERIO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCBr
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (2)