accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COH

CAS RN 112-30-1

IUPAC Name: decan-1-ol
Synonyms: decanol epal 10 1-decanol (natural) n- alcohol decyl royaltac M-2 C9-11 alcohols lorol 22 C 10 alcohol fema number 2365 primary decyl alcohol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: MWKFXSUHUHTGQN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCO
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)