accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-38-9

IUPAC Name: undec-10-enoic acid
Synonyms: desenex 10-undecylenic acid acide 10-undecylique caswell no. 901 ω-hendecenoic acid acido 10-undecenoico renselin hendecenoic acid, ω- cruex 10-undecensaeure
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: FRPZMMHWLSIFAZ-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCC=C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)