accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-39-0

IUPAC Name: methyl hexadecanoate
Synonyms: methyl palmitate
Molecular Weight (g/mol): 270.46
Molecular Formula: C17H34O2
InChi Key: FLIACVVOZYBSBS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)