accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 112-40-3

IUPAC Name: dodecane
Synonyms: dodecane bihexyl dihexyl dodekan adakane 12 ccris 661 N-dodecane
Molecular Weight (g/mol): 170.34
Molecular Formula: C12H26
InChi Key: SNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (4)
  • (11)
Percent Purity 
  • (3)
  • (4)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (10)
Boiling Point 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (7)
  • (4)
  • (4)
  • (11)
  • (3)
  • (4)
  • (7)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (10)
  • (7)
  • (5)