accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-41-4

IUPAC Name: dodec-1-ene
Synonyms: 1-dodecene dodecene (mixture of isomers) N-dodecene
Molecular Weight (g/mol): 168.32
Molecular Formula: C12H24
InChi Key: CRSBERNSMYQZNG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC=C
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (4)
Percent Purity 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)