accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOCH2

CAS RN 112-43-6

IUPAC Name: undec-10-en-1-ol
Synonyms: 10-undecen-1-ol
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: GIEMHYCMBGELGY-UHFFFAOYSA-N
SMILES: OCCCCCCCCCC=C
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)