accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3(E)

CAS RN 112-62-9

IUPAC Name: methyl (9E)-octadec-9-enoate
Synonyms: methyl oleate emery, oleic acid ester oleic acid ester emery Edenor MeTiO5 phytorob 926-67 (E)-methyl oleate (e)-methyl oleate edenor me 90/95V exceparl M-ol emery oleic acid ester 2301
Molecular Weight (g/mol): 296.5
Molecular Formula: C19H36O2
InChi Key: QYDYPVFESGNLHU-ZHACJKMWSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (5)
  • (1)
  • (2)
  • (5)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)