accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3(Z)(Z)

CAS RN 112-63-0

IUPAC Name: methyl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: methyl linoleate
Molecular Weight (g/mol): 294.48
Molecular Formula: C19H34O2
InChi Key: WTTJVINHCBCLGX-NQLNTKRDSA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)