accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-67-4

IUPAC Name: hexadecanoyl chloride
Synonyms: hexadecanoyl chloride
Molecular Weight (g/mol): 274.87
Molecular Formula: C16H31ClO
InChi Key: ARBOVOVUTSQWSS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(Cl)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)