accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 112-71-0

IUPAC Name: 1-bromotetradecane
Synonyms: 1-bromo-N-tetradecane
Molecular Weight (g/mol): 277.29
Molecular Formula: C14H29Br
InChi Key: KOFZTCSTGIWCQG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)