accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOH3C(Z)

CAS RN 112-80-1

IUPAC Name: (9E)-octadec-9-enoic acid
Synonyms: elaidic acid 9-elaidic acid elaidinsaeure
Molecular Weight (g/mol): 282.47
Molecular Formula: C18H34O2
InChi Key: ZQPPMHVWECSIRJ-MDZDMXLPSA-N
SMILES: CCCCCCCC\C=C\CCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (14)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (5)
  • (4)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
Physical Form 
  • (11)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (6)
  • (3)
  • (14)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (4)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (11)
  • (1)
  • (2)