accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-82-3

H3CBr

CAS RN 112-82-3

IUPAC Name: 1-bromohexadecane
Synonyms: 1-bromohexadecane
Molecular Weight (g/mol): 305.34
Molecular Formula: C16H33Br
InChi Key: HNTGIJLWHDPAFN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCBr
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)