accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2

CAS RN 112-88-9

IUPAC Name: octadec-1-ene
Synonyms: octadecene octadecylene α-
Molecular Weight (g/mol): 252.49
Molecular Formula: C18H36
InChi Key: CCCMONHAUSKTEQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCC=C
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results