accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 112-89-0

IUPAC Name: 1-bromooctadecane
Synonyms: 1-bromooctadecane
Molecular Weight (g/mol): 333.4
Molecular Formula: C18H37Br
InChi Key: WSULSMOGMLRGKU-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)
  • (3)
  • (4)