accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNH2(Z)

CAS RN 112-90-3

IUPAC Name: (9Z)-octadec-9-en-1-amine
Synonyms: oleylamine oleamine
Molecular Weight (g/mol): 267.5
Molecular Formula: C18H37N
InChi Key: QGLWBTPVKHMVHM-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCN
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (4)
Physical Form 
  • (4)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (2)