accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112-95-8

IUPAC Name: icosane
Synonyms: eicosane ccris 663
Molecular Weight (g/mol): 282.56
Molecular Formula: C20H42
InChi Key: CBFCDTFDPHXCNY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)