accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 1120-21-4

IUPAC Name: undecane
Synonyms: undecane N-undecane hendekan undekan
Molecular Weight (g/mol): 156.31
Molecular Formula: C11H24
InChi Key: RSJKGSCJYJTIGS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)