accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1120-48-5

IUPAC Name: dioctylamine
Synonyms: 1-octanamine, N-octyl-
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: LAWOZCWGWDVVSG-UHFFFAOYSA-N
SMILES: CCCCCCCCNCCCCCCCC
Boiling Point 
  • (5)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results