accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1120-64-5

IUPAC Name: 2-methyl-4,5-dihydro-1,3-oxazole
Synonyms: 2-methyl-2-oxazoline
Molecular Weight (g/mol): 85.11
Molecular Formula: C4H7NO
InChi Key: GUXJXWKCUUWCLX-UHFFFAOYSA-N
SMILES: CC1=NCCO1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results