CAS RN 1121-89-7
IUPAC Name:
piperidine-2,6-dione
Synonyms:
glutarimide
Molecular Weight (g/mol):
113.12
Molecular Formula:
C5H7NO2
InChi Key:
KNCYXPMJDCCGSJ-UHFFFAOYSA-N
SMILES:
O=C1CCCC(=O)N1
1
–
2
of
2
results
1
Glutarimide, 98%
CAS: 1121-89-7 Molecular Formula: C5H7NO2 Molecular Weight (g/mol): 113.12 MDL Number: MFCD00006670 InChI Key: KNCYXPMJDCCGSJ-UHFFFAOYSA-N Synonym: glutarimide,2,6-piperidinedione,2,6-diketopiperidine,2,6-dioxopiperidine,acetylglycinamide impurity b,piperidin-2,6-dione,acmc-2099da,5-21-09-00557 beilstein handbook reference,2,6-piperidinedione 9ci PubChem CID: 70726 ChEBI: CHEBI:5435 IUPAC Name: piperidine-2,6-dione SMILES: O=C1CCCC(=O)N1