accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112137-89-0

IUPAC Name: 7-[3-hydroxy-2-(4-hydroxy-4-methyloct-1-en-1-yl)-5-oxocyclopentyl]heptanoic acid
Synonyms: 7-[3-hydroxy-2-(4-hydroxy-4-methyloct-1-en-1-yl)-5-oxocyclopentyl]heptanoic acid
Molecular Weight (g/mol): 368.51
Molecular Formula: C21H36O5
InChi Key: CNWGPXZGIIOYDL-UHFFFAOYNA-N
SMILES: CCCCC(C)(O)CC=CC1C(O)CC(=O)C1CCCCCCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)