accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1122-28-7

IUPAC Name: 1H-imidazole-4,5-dicarbonitrile
Synonyms: 1H-imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol): 118.1
Molecular Formula: C5H2N4
InChi Key: XGDRLCRGKUCBQL-UHFFFAOYSA-N
SMILES: N#CC1=C(N=CN1)C#N
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)