accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 112204-58-7

IUPAC Name: 5-bromo-2-fluorophenol
Synonyms: 5-bromo-2-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: YPTHSYKJDRMAJY-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CC=C1F
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results