accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1123-40-6

CH3H3CONHO

CAS RN 1123-40-6

IUPAC Name: 4,4-dimethylpiperidine-2,6-dione
Synonyms: glutarimide, 3,3-dimethyl-
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: YUJCWMGBRDBPDL-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)NC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results