accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1123-56-4

CH3CH3O

CAS RN 1123-56-4

IUPAC Name: 2,6-dimethylbenzaldehyde
Synonyms: 2,6-dimethylbenzaldehyde
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: QOJQBWSZHCKOLL-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1C=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)