accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1123-63-3

CH3ClH3CHO

CAS RN 1123-63-3

IUPAC Name: 4-chloro-2,6-dimethylphenol
Synonyms: phenol, 4-chloro-2,6-dimethyl-
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: VWYKSJIPZHRLNO-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(C)=C1O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)