accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SN

CAS RN 1123-93-9

IUPAC Name: 1,3-benzothiazol-5-amine
Synonyms: 1,3-benzothiazol-5-amine
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: UJZYHMZRXGNDFB-UHFFFAOYSA-N
SMILES: NC1=CC=C2SC=NC2=C1
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)