CAS RN 1124-11-4
IUPAC Name:
tetramethylpyrazine
Synonyms:
tetramethylpyrazine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
FINHMKGKINIASC-UHFFFAOYSA-N
SMILES:
CC1=NC(C)=C(C)N=C1C
1
–
2
of
2
results
1
2,3,5,6-Tetramethylpyrazine, 98+%
CAS: 1124-11-4 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.198 MDL Number: MFCD00006146 InChI Key: FINHMKGKINIASC-UHFFFAOYSA-N Synonym: tetramethylpyrazine,ligustrazine,pyrazine, tetramethyl,bs factor,tetramethylpyrazin,liqustrazine,tetrapyrazine,2,3,5,6-tetramethyl pyrazine,chuanxiongzine,ligustizine PubChem CID: 14296 IUPAC Name: 2,3,5,6-tetramethylpyrazine SMILES: CC1=C(N=C(C(=N1)C)C)C
2
Tetramethylpyrazine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.