accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClHSnH3HClClH

CAS RN 1124-19-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 308.21
Molecular Formula: C6H11Cl3Sn
InChi Key: DQJXDNBAOANFCT-UHFFFAOYSA-N
SMILES: Cl.Cl.Cl.[SnH3]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)